Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
46602-0010
46602-0050
TS46602-0010EA
127
USD
TS46602-0010
TS46602-0050
Dimethyl (1-diazo-2-oxopropyl)phosphonate 95%
Dimethyl (1-diazo-2-oxopropyl)phosphonate
Formula:
C₅H₉N₂O₄P |
MDL Number:
MFCD07368360 CAS Number: 90965-06-3 |
Specification Test Results
Appearance (Color) | Clear light yellow to brown |
Appearance (Form) | Liquid |
Refractive index | 1.4770 to 1.4820 (20°C, 589 nm) |
HPLC | >=94 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...