Benzyl dimethylphosphonoacetate 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dimethylphosphonoacetic Acid Benzyl Ester, Dimethyl (Cbz-methyl)phosphonate, Benzyl 2-(dimethoxyphosphoryl)acetate, Dimethyl (Benzyloxycarbonylmethyl)phosphonate

38737-0050 38737-0250
TS38737-0050EA 165 USD
TS38737-0050 TS38737-0250
Benzyl dimethylphosphonoacetate 98%
Benzyl dimethylphosphonoacetate
Formula: C₁₁H₁₅O₅P
MW: 258.21 g/mol
Boiling Pt: 148…150 °C (.1 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD01872507
CAS Number: 57443-18-2

Order Now

Specification Test Results

Appearance Clear colorless to light brown Liquid
Infrared spectrum Conforms
Refractive index 1.5050 to 1.5070 (20°C, 589 nm)
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR