5,5-Diphenylhydantoin sodium salt 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Phenytoin sodium, Sodium 5,5-Diphenylhydantoin, Sodium 2,4-dioxo-5,5-diphenylimidazolidin-1-ide

43610-0250 43610-1000
TS43610-0250EA 45 USD
TS43610-0250 TS43610-1000
5,5-Diphenylhydantoin sodium salt 99%
5,5-Diphenylhydantoin sodium salt
Formula: C₁₅H₁₂N₂O₂
MW: 274.25 g/mol
Melting Pt: 295…298 °C
Storage Temperature: Ambient
MDL Number: MFCD00069674
CAS Number: 630-93-3

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Water ≤3.0 %
Titration with NaOH 98.5 to 100.5 % (On anhydrous substance)
Color of solution Passes test

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR