3-Ethynylpyridine 96%

Supplier: Thermo Scientific Chemicals
Warning

39807-0010 39807-0050
TS39807-0010EA 66 USD
TS39807-0010 TS39807-0050
3-Ethynylpyridine 96%
3-Ethynylpyridine
Formula: C₇H₅N
Boiling Pt: 83…84 °C (30 mmHg)
Melting Pt: 39…40 °C
Flash Pt: 56 °C
Storage Temperature: Ambient
MDL Number: MFCD02177459
CAS Number: 2510-23-8
UN: 1325
ADR: 4.1,II

Order Now

Specification Test Results

Appearance Colorless to brown Low melting solid (May darken on storage)
Infrared spectrum Conforms
GC ≥95 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR