3-Bromo-2-methyl-1-propene 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-Bromo-2-methylpropene, 2-Methyl-2-propenyl Bromide, 2-Methylallyl Bromide

42765-0050 42765-0250
TS42765-0050EA 82 USD
TS42765-0050 TS42765-0250
3-Bromo-2-methyl-1-propene 97%
3-Bromo-2-methyl-1-propene
Formula: C₄H₇Br
MW: 135.00 g/mol
Boiling Pt: 94…95 °C (760 mmHg)
Density: 1.339 g/cm³
Flash Pt: 7 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00134155
CAS Number: 1458-98-6
UN: 2924
ADR: 3,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4710 to 1.4730 (20°C, 589 nm)
GC ≥96.0 %
Stabilizer Approx 500 ppm hydroquinone

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR