2-Methyl-3-phenyl-2-propen-1-ol 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: alpha-Methylcinnamyl Alcohol, trans-2-Methyl-3-phenyl-2-propen-1-ol

42705-0050 42705-0250
TS42705-0050EA 110 USD
TS42705-0050 TS42705-0250
2-Methyl-3-phenyl-2-propen-1-ol 95%
2-Methyl-3-phenyl-2-propen-1-ol
Formula: C₁₀H₁₂O
MW: 148.21 g/mol
Boiling Pt: 77 °C (.1 mmHg)
Density: 1.03 g/cm³
Flash Pt: 113 °C
Storage Temperature: Ambient
MDL Number: MFCD00004738
CAS Number: 1504-55-8

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.5690 to 1.5710 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR