1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3,3-Dimethyl-1-(trifluoromethyl)-1,2-benziodoxole, Togni Reagent

45690-0010 45690-0050 45690-2500
TS45690-0010EA 223 USD
TS45690-0010 TS45690-0050 TS45690-2500
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole 95%
1-Trifluoromethyl-3,3-dimethyl-1,2-benziodoxole
Formula: C₁₀H₁₀F₃IO
MW: 330.09 g/mol
Melting Pt: 75…79 °C
Storage Temperature: Refrigerator
MDL Number: MFCD10567056
CAS Number: 887144-97-0

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 70 to 79 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR