2-Ethylbenzaldehyde 97%

Supplier: Thermo Scientific Chemicals

46055-0050 46055-0250
TS46055-0050EA 119 USD
TS46055-0050 TS46055-0250
2-Ethylbenzaldehyde 97%
2-Ethylbenzaldehyde
Formula: C₉H₁₀O
MW: 134.1751 g/mol
Boiling Pt: 209 °C
Density: 1.02 g/cm³
Flash Pt: >110 °C
Storage Temperature: Refrigerator
MDL Number: MFCD02261766
CAS Number: 22927-13-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR