3-Oxetanamine 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 3-amino-oxetane, 3-Aminooxetane

45121-2500 45121-5000
TS45121-2500EA 146 USD
TS45121-2500 TS45121-5000
3-Oxetanamine 95%
3-Oxetanamine
Formula: C₃H₇NO
MW: 73.09 g/mol
Boiling Pt: 99 °C
Flash Pt: 24 °C
Storage Temperature: Freezer
MDL Number: MFCD08544363
CAS Number: 21635-88-1
UN: 1992
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR