N-Boc-4-oxo-L-proline 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Boc-4-oxo-Pro-OH, (S)-1-Boc-4-oxo-2-pyrrolidinecarboxylic Acid, (S)-4-Oxopyrrolidine-1,2-dicarboxylic acid 1-tert-butyl ester, (S)-1-(tert-Butoxycarbonyl)-4-oxo-2-pyrrolidinecarboxylic Acid, N-(tert-Butoxycarbonyl)-4-oxo-L-proline

44928-0010 44928-0050
TS44928-0010EA 140 USD
TS44928-0010 TS44928-0050 TS44928-2500
N-Boc-4-oxo-L-proline 98%
N-Boc-4-oxo-L-proline
Formula: C₁₀H₁₅NO₅
MW: 229.23 g/mol
Melting Pt: 155…163 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD01860669
CAS Number: 84348-37-8

Order Now

Specification Test Results

Appearance White to tan Crystalline solid
Infrared spectrum Conforms
Melting point 157 to 163 °C
HPLC ≥97.5 %
Specific optical rotation +18° to +23° (20 °C, 589 nm) (c=0.5, acetone)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR