2-Ethynylpyridine 98+%

Supplier: Thermo Scientific Chemicals

29567-0010 29567-0050 29567-0250
TS29567-0010EA 47 USD
TS29567-0010 TS29567-0050 TS29567-0250
2-Ethynylpyridine 98+%
2-Ethynylpyridine
Formula: C₇H₅N
MW: 103.12 g/mol
Boiling Pt: 77…78 °C (14 mmHg)
Density: 1.02 g/cm³
Flash Pt: 76 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00041598
CAS Number: 1945-84-2

Order Now

Specification Test Results

Appearance Clear dark brown Liquid
Infrared spectrum Conforms
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR