4-(Trifluoromethyl)hydrocinnamic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-[4-(Trifluoromethyl)phenyl]propanoic acid

39738-0010 39738-0050 39738-0250
TS39738-0010EA 48 USD
TS39738-0010 TS39738-0050 TS39738-0250
4-(Trifluoromethyl)hydrocinnamic acid 95%
4-(Trifluoromethyl)hydrocinnamic acid
Formula: C₁₀H₉F₃O₂
MW: 218.17 g/mol
Melting Pt: 106…110 °C
Storage Temperature: Ambient
MDL Number: MFCD00674032
CAS Number: 53473-36-2

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point 111 to 119 °C
HPLC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR