2-Chloro-4-(trifluoromethyl)pyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

39785-0010 39785-0050
TS39785-0010EA 72 USD
TS39785-0010 TS39785-0050
2-Chloro-4-(trifluoromethyl)pyrimidine 97%
2-Chloro-4-(trifluoromethyl)pyrimidine
Formula: C₅H₂ClF₃N₂
MW: 182.5310 g/mol
Boiling Pt: 60 °C (10 mmHg)
Density: 1.513 g/cm³
Flash Pt: 49 °C
Storage Temperature: Ambient
MDL Number: MFCD00115111
CAS Number: 33034-67-2
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR