1H-Pyrazole-1-carboximidamide hydrochloride 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1-Amidinopyrazole Hydrochloride, 1H-Pyrazole-1-carboxamidine monohydrochloride, Hydrochloride, 1-Pyrazolecarboxamidine, Praxadine

36825-0100 36825-0500
TS36825-0100EA 59 USD
TS36825-0100 TS36825-0500
1H-Pyrazole-1-carboximidamide hydrochloride 99%
1H-Pyrazole-1-carboximidamide hydrochloride
Formula: C₄H₇ClN₄
MW: 146.58 g/mol
Melting Pt: 167…170 °C
Storage Temperature: Ambient
MDL Number: MFCD00210087
CAS Number: 4023-02-3

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 167 to 170 °C
Titration Argentometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR