5,7-Dichloro-1,6-naphthyridine 97%

Supplier: Thermo Scientific Chemicals
Warning

44067-0010 44067-0050
TS44067-0010EA 486 USD
TS44067-0010 TS44067-0050
5,7-Dichloro-1,6-naphthyridine 97%
5,7-Dichloro-1,6-naphthyridine
Formula: C₈H₄Cl₂N₂
MW: 199.0368 g/mol
Melting Pt: 138…140 °C
Storage Temperature: Ambient
MDL Number: MFCD11518965
CAS Number: 337958-60-8

Order Now

Specification Test Results

Appearance White to yellow to light orange Crystalline powder or solid
Infrared spectrum Conforms
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR