tert-Butyl-3-oxoazetidine-1-carboxylate 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: N-Boc-3-azetidinone, Tert-butyl 3-oxoazetidine-1-carboxylate, tert-Butyl 3-oxoazetidine-1-carboxylate, 1-BOC-3-Azetidinone

44204-0010 44204-0050
TS44204-0010EA 84 USD
TS44204-0010 TS44204-0050
tert-Butyl-3-oxoazetidine-1-carboxylate 97%
tert-Butyl-3-oxoazetidine-1-carboxylate
Formula: C₈H₁₃NO₃
MW: 171.2 g/mol
Melting Pt: 50…52 °C
Storage Temperature: Ambient
MDL Number: MFCD01861741
CAS Number: 398489-26-4
UN: 3335
ADR: 9,III

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 48 to 54 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR