2-Methylpropane-2-sulphinamide 98%

Supplier: Thermo Scientific Chemicals

42683-0050 42683-0250
TS42683-0050EA 144 USD
TS42683-0050 TS42683-0250
2-Methylpropane-2-sulphinamide 98%
2-Methylpropane-2-sulfinamide
Formula: C₄H₁₁NOS
Melting Pt: 97…101 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01863616
CAS Number: 146374-27-8

Order Now

Specification Test Results

Appearance White to tan Powder or crystals
Infrared spectrum Conforms
GC ≥97.5 %
Specific optical rotation +24° to +31° (20°C, 589 nm) (c=1, toluene)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR