5-Amino-2-chloropyrimidine 95%

Supplier: Thermo Scientific Chemicals
Warning

43432-0010 43432-0050
TS43432-0010EA 178 USD
TS43432-0010 TS43432-0050
5-Amino-2-chloropyrimidine 95%
5-Amino-2-chloropyrimidine
Formula: C₄H₄ClN₃
Melting Pt: 160 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD06412560
CAS Number: 56621-90-0

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder or solid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation +27° to +29° (20°C, 589 nm) (C=4, 5 N HCl)
Titration with HClO4 ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR