Ethyl-6-chloro-3-pyridazinecarboxylate 95%

Supplier: Thermo Scientific Chemicals
Warning

43111-0010 43111-0050
TS43111-0010EA 225 USD
TS43111-0010 TS43111-0050
Ethyl-6-chloro-3-pyridazinecarboxylate 95%
Ethyl-6-chloro-3-pyridazinecarboxylate
Formula: C₇H₇ClN₂O₂
Melting Pt: 149…152 °C
Storage Temperature: Ambient
MDL Number: MFCD09908168
CAS Number: 75680-92-1

Order Now

Specification Test Results

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 148 to 153 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR