4-Iodo-o-anisidine 97%

Supplier: Thermo Scientific Chemicals
Warning

44480-0010 44480-0050
TS44480-0010EA 167 USD
TS44480-0010 TS44480-0050
4-Iodo-o-anisidine 97%
4-Iodo-o-anisidine
Formula: C₇H₈INO
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD11044438
CAS Number: 338454-80-1

Order Now

Specification Test Results

Appearance Brown Solid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR