Biphenyl-4-sulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Biphenyl-4-sulphonyl Chloride

39196-0050 39196-0250
TS39196-0050EA 60 USD
TS39196-0050 TS39196-0250
Biphenyl-4-sulphonyl chloride 97%
Biphenyl-4-sulphonyl chloride
Formula: C₁₂H₉ClO₂S
MW: 252.72 g/mol
Melting Pt: 114 °C
Storage Temperature: Ambient
MDL Number: MFCD00078658
CAS Number: 1623-93-4
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to cream Powder
Infrared spectrum Conforms
Melting point 111 to 116 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR