4-(Chlorosulphonyl)benzoic acid 96%

Supplier: Thermo Scientific Chemicals
Danger

40506-0050 40506-0250
TS40506-0050EA 97 USD
TS40506-0050 TS40506-0250
4-(Chlorosulphonyl)benzoic acid 96%
4-(Chlorosulphonyl)benzoic acid
Formula: C₇H₅ClO₄S
MW: 220.63 g/mol
Melting Pt: 230 °C
Storage Temperature: Ambient
MDL Number: MFCD00007448
CAS Number: 10130-89-9
EINECS: 233-367-2
UN: 3261
ADR: 8 III

Order Now

Specification Test Results

Appearance White to beige Powder or crystals
Infrared spectrum Conforms
Titration excess NaOH ≥95 %
Color scale ≤10 APHA
Free amine ≤0.01 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR