2-(Methylthio)ethylamine 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-(Methylthio)ethanamine

39136-0010 39136-0050
TS39136-0010EA 66 USD
TS39136-0010 TS39136-0050
2-(Methylthio)ethylamine 97%
2-(Methylthio)ethylamine
Formula: C₃H₉NS
MW: 91.18 g/mol
Flash Pt: 36 °C
Storage Temperature: Ambient
MDL Number: MFCD00014825
CAS Number: 18542-42-2
UN: 2734
ADR: 8,II

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4950 to 1.4980 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR