1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 2-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)acetate

44501-0010 44501-0050
TS44501-0010EA 118 USD
TS44501-0010 TS44501-0050
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester 97%
1-(Ethoxycarbonylmethyl)-1H-pyrazole-4-boronic acid pinacol ester
Formula: C₁₃H₂₁BN₂O₄
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD08706026
CAS Number: 864754-16-5

Order Now

Specification Test Results

Appearance Colorless to yellow to orange Viscous liquid
Infrared spectrum Conforms
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR