3-Amino-1-bromoisoquinoline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1-Bromoisoquinolin-3-amine

44916-0010 44916-0050
TS44916-0010EA 170 USD
TS44916-0010 TS44916-0050
3-Amino-1-bromoisoquinoline 97%
3-Amino-1-bromoisoquinoline
Formula: C₉H₇BrN₂
MW: 223.07 g/mol
Melting Pt: 144…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00102193
CAS Number: 13130-79-5

Order Now

Specification Test Results

Appearance Yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 147 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR