(4-Methylphenyl)methanesulphonyl chloride 97%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: p-Tolylmethanesulphonyl chloride, α-p-Xylenesulphonyl chloride, 4-Methylbenzylsulphonyl chloride

45484-0010 45484-0050
TS45484-0010EA 76 USD
TS45484-0010 TS45484-0050
(4-Methylphenyl)methanesulphonyl chloride 97%
(4-Methylphenyl)methanesulphonyl chloride
Formula: C₈H₉ClO₂S
MW: 204.67 g/mol
Melting Pt: 78…82 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01631931
CAS Number: 51419-59-1
UN: 3261
ADR: 8,II

Order Now

Specification Test Results

Appearance White to tan Powder or solid
Infrared spectrum Conforms
Melting point 75 to 81 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR