(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) 98+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 1H-Benzotriazol-1-yloxytris(dimethylamino)phosphonium hexafluorophosphate, ((1H-Benzo[d][1,2,3]triazol-1-yl)oxy)tris(dimethylamino)phosphonium hexafluorophosphate(V), BOP reagent, Benzotriazol-1-yloxytris(dimethylamino)-phosphonium hexafluorophosphate

20980-0010 20980-0050 20980-0250
TS20980-0010EA 36 USD
TS20980-0010 TS20980-0050 TS20980-0250
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP) 98+%
(Benzotriazol-1-yloxy)tris(dimethylamino)phosphonium hexafluorophosphate (BOP)
Formula: C₁₂H₂₂F₆N₆OP₂
MW: 442.28 g/mol
Melting Pt: 136…140 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011948
CAS Number: 56602-33-6
EINECS: 260-279-1
UN: 1325

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 136 to 140 °C
HPLC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR