2,2,3,3,4,4,4-Heptafluorobutylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
36877-0010
36877-0050
TS36877-0010EA
68
USD
TS36877-0010
TS36877-0050
2,2,3,3,4,4,4-Heptafluorobutylamine 97%
2,2,3,3,4,4,4-Heptafluorobutylamine
Formula:
C₄H₄F₇N MW: 199.07 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00014817 CAS Number: 374-99-2 EINECS: 206-780-0 UN: 2735 ADR: 8,III |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...