4-Iodo-1-methyl-1H-pyrazole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
37179-0010
37179-0050
TS37179-0010EA
64
USD
TS37179-0010
TS37179-0050
4-Iodo-1-methyl-1H-pyrazole 97%
4-Iodo-1-methyl-1H-pyrazole
Formula:
C₄H₅IN₂ MW: 208.0000 g/mol Melting Pt: 61…67 °C Storage Temperature: Ambient |
MDL Number:
MFCD05663860 CAS Number: 39806-90-1 |
Specification Test Results
Appearance | Off-white Semitransparent crystals or crystalline needles |
Infrared spectrum | Conforms |
Melting point | 61 to 67 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...