3-Iodo-1H-pyrazole 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Iodopyrazole

44481-0010 44481-0050
TS44481-0010EA 143 USD
TS44481-0010 TS44481-0050
3-Iodo-1H-pyrazole 97%
3-Iodo-1H-pyrazole
Formula: C₃H₃IN₂
MW: 193.98 g/mol
Melting Pt: 68…73 °C
Storage Temperature: Ambient
MDL Number: MFCD12022324
CAS Number: 4522-35-4

Order Now

Specification Test Results

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 67 to 74 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR