2-Bromo-2',4'-difluoroacetophenone 95%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-1-(2,4-difluorophenyl)ethanone, 2,4-Difluorophenacyl bromide

44228-0010 44228-0050
TS44228-0010EA 85 USD
TS44228-0010 TS44228-0050
2-Bromo-2',4'-difluoroacetophenone 95%
2-Bromo-2',4'-difluoroacetophenone
Formula: C₈H₅BrF₂O
MW: 235.03 g/mol
Melting Pt: 31…33 °C
Flash Pt: 51 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00142822
CAS Number: 102429-07-2
UN: 2925
ADR: 4.1,II

Order Now

Specification Test Results

Appearance White to light yellow Low melting solid
Infrared spectrum Conforms
Melting point 30 to 35 °C
GC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR