N,N-Bis(trifluoromethanesulphonyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 1,1,1-Trifluoro-N-phenyl-N-((trifluoromethyl)sulphonyl)methanesulphonamide, N-Phenylbis(trifluoromethanesulphonimide)

37479-0050 37479-0250 37479-1000
TS37479-0050EA 75 USD
TS37479-0050 TS37479-0250 TS37479-1000
N,N-Bis(trifluoromethanesulphonyl)aniline 97%
N,N-Bis(trifluoromethanesulphonyl)aniline
Formula: C₈H₅F₆NO₄S₂
MW: 357.25 g/mol
Melting Pt: 99…105 °C
Storage Temperature: Ambient
MDL Number: MFCD00000404
CAS Number: 37595-74-7

Order Now

Specification Test Results

Appearance White to yellow to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 99 to 105 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR