2-(Trifluoromethyl)acrylic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

31310-0010 31310-0050
TS31310-0010EA 64 USD
TS31310-0010 TS31310-0050
2-(Trifluoromethyl)acrylic acid 98%
2-(Trifluoromethyl)acrylic acid
Formula: C₄H₃F₃O₂
Boiling Pt: 86 °C (760 mmHg)
Melting Pt: 48…52 °C
Flash Pt: 81 °C
Storage Temperature: Ambient
MDL Number: MFCD00042424
CAS Number: 381-98-6

Order Now

Specification Test Results

Appearance Transparent light yellow Chunks
Infrared spectrum Conforms
Melting point 48 to 52 °C
Titration with TBAH ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR