2,3-Difluoroisonicotinic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44677-0010
44677-0050
TS44677-0010EA
109
USD
TS44677-0010
TS44677-0050
2,3-Difluoroisonicotinic acid 97%
2,3-Difluoroisonicotinic acid
Formula:
C₆H₃F₂NO₂ Melting Pt: 165…167 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD09031174 CAS Number: 851386-31-7 |
Specification Test Results
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
Titration with NaOH | ≥96.0 to ≤104.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...