Salicylaldehyde azine 99%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Bis(2-hydroxybenzaldehyde) hydrazone, N,N'-Bis(salicylidene)hydrazine, Salazine, Salicylaldazine, 2,2'-(Hydrazine-1,2-diylidenebis(methanylylidene))diphenol

45694-1000 45694-0050 45694-0250
TS45694-1000EA 726 USD
TS45694-1000 TS45694-0050 TS45694-0250
Salicylaldehyde azine 99%
Salicylaldehyde azine
Formula: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Melting Pt: 218…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00043496
CAS Number: 959-36-4

Order Now

Specification Test Results

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR