2-Methoxy-4-pyridineboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Methoxy-4-pyridylboronic acid, 2-Methoxypyridine-4-boronic acid, (2-Methoxypyridin-4-yl)boronic acid

45437-0010 45437-2500
TS45437-0010EA 488 USD
TS45437-0010 TS45437-2500
2-Methoxy-4-pyridineboronic acid 95%
2-Methoxy-4-pyridineboronic acid
Formula: C₆H₈BNO₃
MW: 152.9436 g/mol
Melting Pt: 65…70 °C
Storage Temperature: Refrigerator
MDL Number: MFCD07368877
CAS Number: 762262-09-9

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR