Diethyl (hydroxymethyl)phosphonate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
32095-0050
32095-0250
TS32095-0050EA
62
USD
TS32095-0050
TS32095-0250
Diethyl (hydroxymethyl)phosphonate 97%
Diethyl (hydroxymethyl)phosphonate
Formula:
C₅H₁₃O₄P MW: 168.13 g/mol Boiling Pt: 124…126 °C (3 mmHg) Density: 1.14 g/cm³ Flash Pt: >110 °C Storage Temperature: Ambient |
MDL Number:
MFCD00014418 CAS Number: 3084-40-0 EINECS: 221-391-6 |
Specification Test Results
Appearance | Clear colorless to yellow Viscous liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...