2,2-Dimethylcyclohexanone

Supplier: Thermo Scientific Chemicals
Warning

42650-0010 42650-0050
TS42650-0010EA 274 USD
TS42650-0010 TS42650-0050
2,2-Dimethylcyclohexanone
2,2-Dimethylcyclohexanone
Formula: C₈H₁₄O
MW: 126.1962 g/mol
Boiling Pt: 169…170 °C (.8 mmHg)
Melting Pt: –20 °C
Density: 0.912 g/cm³
Flash Pt: 50 °C
Storage Temperature: Ambient
MDL Number: MFCD00134382
CAS Number: 1193-47-1
UN: 1224
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4470 to 1.4490 (20°C, 589 nm)
GC ≥94 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR