Di-tert-butyl phosphonate 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Di-tert-butyl phosphite
44939-0010
44939-0050
44939-0250
TS44939-0010EA
31
USD
TS44939-0010
TS44939-0050
TS44939-0250
Di-tert-butyl phosphonate 96%
Di-tert-butyl phosphonate
Formula:
C₈H₁₉O₃P MW: 194.21 Boiling Pt: 66…68 °C (.5 mmHg) Flash Pt: 163 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00995019 CAS Number: 13086-84-5 |
Specification Test Results
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4190 to 1.4210 (20°C, 589 nm) |
GC | ≥95.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...