Dibenzyl diisopropylphosphoramidite 90+%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Dibenzyl N,N-diisopropylphosphoramidite

39964-0250 39964-0050
TS39964-0250EA 306 USD
TS39964-0250 TS39964-0050
Dibenzyl diisopropylphosphoramidite 90+%
Dibenzyl diisopropylphosphoramidite
Formula: C₂₀H₂₈NO₂P
MW: 345.43 g/mol
Density: 1.028 g/cm³
Flash Pt: 69 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00191988
CAS Number: 108549-23-1

Order Now

Specification Test Results

Appearance Clear to hazy colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR