(Methyl)triphenoxyphosphonium iodide 95%

Supplier: Thermo Scientific Chemicals

41502-0050 41502-0500
TS41502-0050EA 44 USD
TS41502-0050 TS41502-0500
(Methyl)triphenoxyphosphonium iodide 95%
(Methyl)triphenoxyphosphonium iodide
Formula: C₁₉H₁₈IO₃P
Melting Pt: 141…146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011911
CAS Number: 17579-99-6
EINECS: 241-551-9

Order Now

Specification Test Results

Appearance Orange to brown Moist powder or crystals
Infrared spectrum Conforms
Titration Argentometric ≥94 %
GC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR