Perfluorobutane-1-sulphonyl fluoride 92%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
Perfluoro-1-butanesulphonyl fluoride, Nonafluorobutanesulphonyl fluoride
31234-0050
31234-0250
31234-1000
TS31234-0050EA
29
USD
TS31234-0050
TS31234-0250
TS31234-1000
Perfluorobutane-1-sulphonyl fluoride 92%
Perfluorobutane-1-sulphonyl fluoride
Formula:
C₄F₁₀O₂S MW: 302.09 Boiling Pt: 64 °C Density: 1.68 g/cm³ Storage Temperature: Ambient |
MDL Number:
MFCD00007422 CAS Number: 375-72-4 EINECS: 206-792-6 UN: 3265 ADR: 8,II |
Specification Test Results
Appearance | Clear colorless Liquid |
Infrared spectrum | Conforms |
GC | ≥90 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...