Octyl-β-D-thioglucopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Octyl-thioglucoside, OTG, 1-S-Octyl-β-D-thioglucopyranoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-(octylthio)tetrahydro-2H-pyran-3,4,5-triol, N-Octyl-1-thio-Beta-D-glucopyranoside, Octyl β-D-thioglucopyranoside
31500-0025
31500-0100
TS31500-0025EA
180
USD
TS31500-0025
TS31500-0100
TS31500-5000
Octyl-β-D-thioglucopyranoside 98%
Octyl β-D-thioglucopyranoside
Formula:
C₁₄H₂₈O₅S MW: 308.44 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD00012189 CAS Number: 85618-21-9 |
Specification Test Results
Appearance | White Crystalline powder and/or chunks |
Infrared spectrum | Conforms |
HPLC | ≥97.5 % (ELSD) |
Specific optical rotation | -52° to -54° (20°C, 589 nm) (c=1, CH3OH) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...