Diphosphoryl tetrachloride 90+%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Diphosphoryl chloride, Pyrophosphoryl chloride

39362-0050 39362-0250
TS39362-0050EA 70 USD
TS39362-0050 TS39362-0250
Diphosphoryl tetrachloride 90+%
Diphosphoryl tetrachloride
Formula: Cl₄O₃P₂
MW: 251.76 g/mol
Boiling Pt: 90 °C (12 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00061542
CAS Number: 13498-14-1
UN: 3264
ADR: 8,I

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid
Infrared spectrum Conforms
Titration with NaOH ≥90.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR