(R)-(+)-4-Isopropyl-2-oxazolidinone 98%

Supplier: Thermo Scientific Chemicals

Synonyms: (R)-4-Isopropyloxazolidin-2-one, (4R)-(+)-4-Isopropyl-2-oxazolidinone

29785-0010 29785-0050 29785-0250
TS29785-0010EA 64 USD
TS29785-0010 TS29785-0050 TS29785-0250
(R)-(+)-4-Isopropyl-2-oxazolidinone 98%
(R)-(+)-4-Isopropyl-2-oxazolidinone
Formula: C₆H₁₁NO₂
MW: 129.16 g/mol
Melting Pt: 70…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00075172
CAS Number: 95530-58-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Melting point 70 to 74 °C
GC ≥97.5 %
Specific optical rotation +16° to +20° (20 °C, 589 nm) (c=1, ethanol)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR