2-Chloro-4,6-dimethylpyrimidine 97%

Supplier: Thermo Scientific Chemicals
Warning

44255-0050 44255-0250
TS44255-0050EA 94 USD
TS44255-0050 TS44255-0250
2-Chloro-4,6-dimethylpyrimidine 97%
2-Chloro-4,6-dimethylpyrimidine
Formula: C₆H₇ClN₂
MW: 142.59 g/mol
Boiling Pt: 210…212 °C (760 mmHg)
Melting Pt: 36…38 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00023199
CAS Number: 4472-44-0

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder or low melting solid
Infrared spectrum Conforms
Melting point 34 to 39 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR