Methyl phenyl sulphoxide 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: (Methylsulphinyl)benzene

35212-0050 35212-0250
TS35212-0050EA 50 USD
TS35212-0050 TS35212-0250
Methyl phenyl sulphoxide 98+%
Methyl phenyl sulphoxide
Formula: C₇H₈OS
MW: 140.21 g/mol
Boiling Pt: 92…94 °C (.3 mmHg)
Melting Pt: 34…36 °C
Flash Pt: 86 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00002088
CAS Number: 1193-82-4
EINECS: 214-781-2

Order Now

Specification Test Results

Appearance Clear colorless to yellow Liquid or fused mass
Infrared spectrum Conforms
Refractive index 1.5755 to 1.5795 (20°C, 589 nm)
GC ≥98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR