2,5-Dimethyl-p-benzoquinone 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2,5-Dimethylcyclohexa-2,5-diene-1,4-dione

40823-0050 40823-0250
TS40823-0050EA 135 USD
TS40823-0050 TS40823-0250
2,5-Dimethyl-p-benzoquinone 99%
2,5-Dimethyl-p-benzoquinone
Formula: C₈H₈O₂
MW: 136.15 g/mol
Melting Pt: 123…126 °C
Storage Temperature: Ambient
MDL Number: MFCD00041737
CAS Number: 137-18-8
EINECS: 205-283-6
UN: 2811

Order Now

Specification Test Results

Appearance Yellow to brown Crystalline powder
Infrared spectrum Conforms
Melting point 123 to 126 °C
GC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR