2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
44220-0010
44220-0050
TS44220-0010EA
56
USD
TS44220-0010
TS44220-0050
2,3-Dimethoxy-5-methyl-p-benzoquinone 98%
2,3-Dimethoxy-5-methyl-p-benzoquinone
Formula:
C₉H₁₀O₄ Melting Pt: 59…61 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00001595 CAS Number: 605-94-7 |
Specification Test Results
Appearance | Orange to red Crystalline powder or needles |
Infrared spectrum | Conforms |
Melting point | 58 to 62 °C |
HPLC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...