2,6-Dimethoxy-p-benzoquinone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,6-Dimethoxy-1,4-Benzoquinone
33123-0050
33123-0250
TS33123-0050EA
62
USD
TS33123-0050
TS33123-0250
2,6-Dimethoxy-p-benzoquinone 97%
2,6-Dimethoxy-p-benzoquinone
Formula:
C₈H₈O₄ MW: 168.15 g/mol Melting Pt: 253…257 °C Storage Temperature: Ambient |
MDL Number:
MFCD00016368 CAS Number: 530-55-2 EINECS: 208-484-7 |
Specification Test Results
Appearance | Yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 253 to 257 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...